Assay Details
Target Gene Details
Entrez Gene ID: | 52552 |
Gene Name: | poly (ADP-ribose) polymerase family, member 8 |
Gene Aliases: |
2810430O08Rik, ARTD16, D13Ertd275e |
Location: |
Chr.13:116850842-117025913 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp8 | NM_001081009.1 | NP_001074478.1 | ||
| AK149889.1 | 1 | 133 | ||
| AK149918.1 | 1 | 133 | BAE29166.1 | |
| AK150113.1 | 1 | 79 | BAE29317.1 | |
| AK150208.1 | 1 | 133 | BAE29379.1 | |
| AK150732.1 | ||||
| AK151510.1 | ||||
| AK152919.1 | ||||
| AK153169.1 | ||||
| AK153270.1 | ||||
| AK155693.1 | BAE33392.1 | |||
| BC021881.1 | 1 | 136 | AAH21881.1 |

Genomic Map