Assay Details
Target Gene Details
Entrez Gene ID: | 52552 |
Gene Name: | poly (ADP-ribose) polymerase family, member 8 |
Gene Aliases: |
2810430O08Rik, ARTD16, D13Ertd275e |
Location: |
Chr.13:116850842-117025913 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp8 | NM_001081009.1 | 24 | 2430 | NP_001074478.1 |
| AK013206.1 | 8 | 582 | BAB28711.2 | |
| AK149889.1 | 23 | 2403 | ||
| AK149918.1 | 23 | 2397 | BAE29166.1 | |
| AK150208.1 | 23 | 2397 | BAE29379.1 | |
| AK150732.1 | 22 | 2207 | ||
| AK151510.1 | 22 | 2208 | ||
| AK152919.1 | 22 | 2207 | ||
| AK153169.1 | 22 | 2207 | ||
| AK153270.1 | 22 | 2207 | ||
| BC021315.1 | 14 | 1564 | AAH21315.1 | |
| BC021881.1 | 23 | 2400 | AAH21881.1 | |
| BC022679.1 | 9 | 957 | AAH22679.2 |

Genomic Map