Assay Details
Target Gene Details
Entrez Gene ID: | 97863 |
Gene Name: | expressed sequence C78339 |
Gene Aliases: |
- |
Location: |
Chr.13:46669443-46678064 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C78339 | NM_001033192.2 | 3 | 908 | NP_001028364.1 |
| AK090371.1 | 3 | 292 | ||
| AK141282.1 | 3 | 909 | BAE24634.1 | |
| AK171039.1 | 4 | 809 | BAE42204.1 | |
| AK218359.1 | 2 | 187 |

Genomic Map