Assay Details
Target Gene Details
Entrez Gene ID: | 14569 |
Gene Name: | guanosine diphosphate (GDP) dissociation inhibitor 2 |
Gene Aliases: |
GDI-B, GDIB, Gdi3 |
Location: |
Chr.13:3538075-3566261 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdi2 | NM_008112.4 | NP_032138.3 | ||
| AK007871.1 | BAB25321.1 | |||
| AK013104.1 | BAB28649.1 | |||
| AK078514.1 | ||||
| AK079690.1 | BAC37725.1 | |||
| AK083063.1 | ||||
| AK090083.1 | BAC41085.1 | |||
| AK137761.1 | BAE23494.1 | |||
| AK143633.1 | BAE25473.1 | |||
| AK143681.1 | BAE25494.1 | |||
| AK149958.1 | BAE29194.1 | |||
| AK150356.1 | ||||
| AK150655.1 | BAE29742.1 | |||
| AK152250.1 | BAE31072.1 | |||
| AK153381.1 | BAE31947.1 | |||
| AK159734.1 | BAE35328.1 | |||
| AK166317.1 | BAE38700.1 | |||
| AK167659.1 | BAE39709.1 | |||
| AK169299.1 | BAE41055.1 | |||
| AK186127.1 | ||||
| BC055341.1 | AAH55341.1 | |||
| L36314.1 | AAA78786.1 |
More Information
Set Membership: |
|

Genomic Map