Assay Details
Target Gene Details
Entrez Gene ID: | 16563 |
Gene Name: | kinesin family member 2A |
Gene Aliases: |
C530030B14Rik, Kif2, Kns2, M-kinesin |
Location: |
Chr.13:106960585-107022228 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif2a | NM_001145779.1 | NP_001139251.1 | ||
| NM_008442.2 | NP_032468.2 | |||
| AK037037.1 | ||||
| BC006803.1 | ||||
| D12644.1 | BAA02165.1 |
More Information
Set Membership: |
|

Genomic Map