Assay Details
Target Gene Details
Entrez Gene ID: | 140580 |
Gene Name: | engulfment and cell motility 1 |
Gene Aliases: |
6330578D22Rik, C230095H21Rik, CED-12 |
Location: |
Chr.13:20090240-20608353 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elmo1 | NM_080288.2 | 8 | 841 | NP_525027.1 |
| AF398883.1 | 7 | 505 | AAL14464.1 | |
| AK028389.1 | 8 | 916 | BAC25925.1 | |
| AK220253.1 | 8 | 729 | BAD90178.1 |
More Information
Set Membership: |
|

Genomic Map