Assay Details
Target Gene Details
Entrez Gene ID: | 73942 |
Gene Name: | family with sequence similarity 151, member B |
Gene Aliases: |
4930405M20Rik |
Location: |
Chr.13:92449620-92484290 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam151b | NM_001163627.1 | NP_001157099.1 | ||
| AK015100.1 | ||||
| AK162422.1 |

Genomic Map