Assay Details
Target Gene Details
Entrez Gene ID: | 544963 |
Gene Name: | IQ motif containing GTPase activating protein 2 |
Gene Aliases: |
4933417J23Rik, A630053O10, AI788777 |
Location: |
Chr.13:95627177-95891969 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqgap2 | NM_027711.1 | 16 | 2033 | NP_081987.1 |
| AK142813.1 | 16 | 2034 | BAE25199.1 | |
| AK147360.1 | 16 | 2027 | ||
| BC052916.1 | 16 | 1873 | AAH52916.1 | |
| BC094939.1 | 16 | 1861 | AAH94939.1 | |
| BC150958.1 | 16 | 2032 |
More Information
Set Membership: |
|

Genomic Map