Assay Details
Target Gene Details
Entrez Gene ID: | 100503085 |
Gene Name: | kelch-like 3 |
Gene Aliases: |
7530408C15Rik, AI430941, EG627648 |
Location: |
Chr.13:58004439-58123481 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl3 | NM_001195075.1 | 17 | 2022 | NP_001182004.1 |
More Information
Set Membership: |
|

Genomic Map