Assay Details
Target Gene Details
Entrez Gene ID: | 12626 |
Gene Name: | centrin 3 |
Gene Aliases: |
MmCEN3 |
Location: |
Chr.13:81783247-81797157 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cetn3 | NM_007684.3 | NP_031710.1 | ||
| AK003473.1 | ||||
| AK004532.1 | BAB23351.1 | |||
| AK006289.1 | BAB24508.1 | |||
| AK006891.1 | BAB24781.1 | |||
| AK011821.1 | BAB27862.1 | |||
| AK017505.1 | BAB30778.1 | |||
| BC002162.1 | AAH02162.1 | |||
| BC054097.1 | AAH54097.1 | |||
| Y12474.1 | CAA73078.1 |
More Information
Set Membership: |
|

Genomic Map