Assay Details
Target Gene Details
Entrez Gene ID: | 74559 |
Gene Name: | ELOVL family member 7, elongation of long chain fatty acids (yeast) |
Gene Aliases: |
9130013K24Rik, AI840082 |
Location: |
Chr.13:108214328-108287109 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl7 | NM_029001.5 | 3 | 325 | NP_083277.3 |
| AK018616.1 | 3 | 300 | BAB31310.1 | |
| AK037162.1 | 3 | 297 | BAC29727.1 | |
| AK049118.1 | 3 | 325 | BAC33552.1 | |
| AK050441.1 | 3 | 320 | ||
| AK137668.1 | 3 | 310 | BAE23454.1 | |
| BC005602.1 | 3 | 325 | AAH05602.1 |
More Information
Set Membership: |
|

Genomic Map