Assay Details
Target Gene Details
Entrez Gene ID: | 193385 |
Gene Name: | family with sequence similarity 65, member B |
Gene Aliases: |
- |
Location: |
Chr.13:24501534-24733806 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam65b | NM_001286100.1 | 15 | 2100 | NP_001273029.1 |
| NM_029679.2 | 15 | 2232 | NP_083955.1 | |
| AK122269.1 | 15 | 2164 | BAC65551.1 | |
| AK134538.1 | 15 | 2162 |
More Information
Set Membership: |
|

Genomic Map