Assay Details
Target Gene Details
Entrez Gene ID: | 26913 |
Gene Name: | G protein-regulated inducer of neurite outgrowth 1 |
Gene Aliases: |
GRIN1, Z16 |
Location: |
Chr.13:54736671-54750150 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprin1 | NM_012014.3 | NP_036144.2 | ||
| AK138306.1 | BAE23617.1 | |||
| AK144210.1 | BAE25773.1 | |||
| BC057044.1 | AAH57044.1 |

Genomic Map