Assay Details
Target Gene Details
Entrez Gene ID: | 28126 |
Gene Name: | NOP16 nucleolar protein |
Gene Aliases: |
AA409471, D13Wsu177e |
Location: |
Chr.13:54584191-54590074 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nop16 | NM_178605.4 | NP_848720.1 | ||
| AK002696.1 | BAB22290.1 | |||
| AK003769.1 | ||||
| AK013251.1 | BAB28746.1 | |||
| AK088248.1 | BAC40235.1 | |||
| BC012213.1 | AAH12213.1 |

Genomic Map