Assay Details
Target Gene Details
Entrez Gene ID: | 18548 |
Gene Name: | proprotein convertase subtilisin/kexin type 1 |
Gene Aliases: |
Nec-1, Nec1, PC1, PC3, Phpp-1, SPC3 |
Location: |
Chr.13:75089415-75134861 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcsk1 | NM_013628.2 | 11 | 1546 | NP_038656.1 |
| BC108982.1 | 11 | 1524 | AAI08983.1 | |
| BC108983.1 | 11 | 1524 | AAI08984.1 | |
| M58589.1 | 11 | 1546 | AAA39894.1 | |
| M69196.1 | 11 | 1652 | AAA39732.1 | |
| X57088.1 | 11 | 1658 | CAA40368.1 |
More Information
Set Membership: |
|

Genomic Map