Assay Details
Target Gene Details
Entrez Gene ID: | 69635 |
Gene Name: | death associated protein kinase 1 |
Gene Aliases: |
D13Ucla1, DAP-Kinase |
Location: |
Chr.13:60601947-60763191 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dapk1 | NM_001285917.1 | 10 | 1145 | NP_001272846.1 |
| NM_029653.3 | 10 | 1145 | NP_083929.2 | |
| NM_134062.2 | 10 | 1030 | NP_598823.1 | |
| AK009701.1 | 10 | 1035 | ||
| AK145922.1 | 10 | 1031 | BAE26753.1 | |
| AY245540.2 | 10 | 1138 | AAO91934.2 | |
| AY245541.1 | 10 | 1131 | AAO91935.1 | |
| BC021490.1 | 8 | 629 | AAH21490.1 | |
| X97048.1 | 9 | 980 | CAA65762.1 |
More Information
Set Membership: |
|

Genomic Map