Assay Details
Target Gene Details
Entrez Gene ID: | 69635 |
Gene Name: | death associated protein kinase 1 |
Gene Aliases: |
D13Ucla1, DAP-Kinase |
Location: |
Chr.13:60601947-60763191 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dapk1 | NM_001285917.1 | NP_001272846.1 | ||
| NM_029653.3 | NP_083929.2 | |||
| NM_134062.2 | NP_598823.1 | |||
| AK145922.1 | BAE26753.1 | |||
| AY245540.2 | AAO91934.2 | |||
| AY245541.1 | AAO91935.1 | |||
| BC021490.1 | AAH21490.1 | |||
| X97048.1 | CAA65762.1 |
More Information
Set Membership: |
|

Genomic Map