Assay Details
Target Gene Details
Entrez Gene ID: | 67044 |
Gene Name: | HIG1 domain family, member 2A |
Gene Aliases: |
2010110M21Rik, AA690237 |
Location: |
Chr.13:54590231-54591147 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Higd2a | NM_025933.3 | NP_080209.1 | ||
| AK004272.1 | ||||
| AK005269.1 | BAB23921.1 | |||
| AK008387.1 | BAB25641.1 | |||
| BC021471.1 | AAH21471.1 |

Genomic Map