Assay Details
Target Gene Details
Entrez Gene ID: | 74325 |
Gene Name: | clathrin, light polypeptide (Lcb) |
Gene Aliases: |
2310046E19Rik |
Location: |
Chr.13:54592939-54611397 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cltb | NM_028870.3 | NP_083146.1 | ||
| AK029382.1 | BAC26430.1 | |||
| AK045989.1 | BAC32563.1 | |||
| AK159480.1 | BAE35117.1 | |||
| BC070404.1 | AAH70404.1 |
More Information
Set Membership: |
|

Genomic Map