Assay Details
Target Gene Details
Entrez Gene ID: | 193385 |
Gene Name: | family with sequence similarity 65, member B |
Gene Aliases: |
- |
Location: |
Chr.13:24501534-24733806 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam65b | NM_001080381.1 | NP_001073850.1 | ||
| NM_001286100.1 | NP_001273029.1 | |||
| NM_001286101.1 | NP_001273030.1 | |||
| NM_029679.2 | NP_083955.1 | |||
| NM_178658.5 | NP_848773.3 | |||
| AK031928.1 | BAC27606.1 | |||
| AK036452.1 | BAC29436.1 | |||
| AK039271.1 | BAC30301.1 | |||
| AK040887.1 | BAC30732.1 | |||
| AK041665.1 | BAC31025.1 | |||
| AK122269.1 | BAC65551.1 | |||
| AK134538.1 | ||||
| AK163880.1 | BAE37527.1 | |||
| BB638362.1 |
More Information
Set Membership: |
|

Genomic Map