Assay Details
Target Gene Details
Entrez Gene ID: | 212032 |
Gene Name: | hexokinase 3 |
Gene Aliases: |
HK III, HK-III |
Location: |
Chr.13:55005985-55021472 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hk3 | NM_001033245.4 | NP_001028417.1 | ||
| NM_001206390.1 | NP_001193319.1 | |||
| NM_001206391.1 | NP_001193320.1 | |||
| NM_001206392.1 | NP_001193321.1 | |||
| AK140444.1 | BAE24390.1 | |||
| AK149651.1 | BAE29007.1 | |||
| AK149992.1 | BAE29220.1 | |||
| AK154603.1 | BAE32706.1 | |||
| AK155701.1 | ||||
| AK159857.1 | ||||
| AK159906.1 | BAE35470.1 | |||
| AK160018.1 | BAE35564.1 | |||
| AK160063.1 | BAE35599.1 | |||
| AK162637.1 | BAE37000.1 | |||
| AK163658.1 | BAE37444.1 | |||
| AK170813.1 | BAE42045.1 | |||
| AK171214.1 | BAE42318.1 | |||
| AK171248.1 | BAE42341.1 | |||
| AK171580.1 | BAE42538.1 | |||
| BC117860.1 | ||||
| BC117861.1 |
More Information
Set Membership: |
|

Genomic Map