Assay Details
Target Gene Details
Entrez Gene ID: | 76041 |
Gene Name: | coiled-coil domain containing 125 |
Gene Aliases: |
5830436D01Rik, AL023012 |
Location: |
Chr.13:100668934-100697240 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc125 | NM_001168386.2 | 8 | 1144 | NP_001161858.1 |
| NM_183115.5 | 9 | 1222 | NP_898938.4 | |
| AK017973.1 | 8 | 907 | BAC25536.1 | |
| BC085249.1 | 9 | 940 | AAH85249.1 |

Genomic Map