Assay Details
Target Gene Details
Entrez Gene ID: | 14088 |
Gene Name: | Fanconi anemia, complementation group C |
Gene Aliases: |
Facc, mir-3074-1 |
Location: |
Chr.13:63301134-63497290 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fancc | NM_001042673.2 | 2 | 437 | NP_001036138.1 |
| NM_001282942.1 | NP_001269871.1 | |||
| NM_007985.3 | 2 | 437 | NP_032011.2 | |
| AK035488.1 | 2 | 191 | BAC29076.1 | |
| AK042226.1 | 2 | 405 | ||
| AK082717.1 | BAE43379.1 | |||
| AK147025.1 | 2 | 417 | BAE27618.1 | |
| AK163161.1 | 2 | 438 | ||
| AK196042.1 | 2 | 167 | ||
| BC038609.1 | AAH38609.1 | |||
| L08266.1 | 2 | 418 | AAA37590.1 |

Genomic Map