Assay Details
Target Gene Details
Entrez Gene ID: | 105278 |
Gene Name: | cyclin-dependent kinase 20 |
Gene Aliases: |
4932702G04Rik, AU019450, AW558027, CDCH, Ccrk, PNQLARE, p42 |
Location: |
Chr.13:64432363-64439722 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk20 | NM_053180.2 | 6 | 662 | NP_444410.1 |
| AY005133.1 | 6 | 673 | AAF89089.1 | |
| AY904369.1 | AAW82351.1 | |||
| BC031907.1 | 6 | 662 | AAH31907.1 |
More Information
Set Membership: |
|

Genomic Map