Assay Details
Target Gene Details
Entrez Gene ID: | 14120 |
Gene Name: | fructose bisphosphatase 2 |
Gene Aliases: |
Fbp-1, Fbp1, Rae-30 |
Location: |
Chr.13:62836885-62858370 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbp2 | NM_007994.3 | 2 | 323 | NP_032020.2 |
| AJ132692.1 | 2 | 250 | CAB65243.1 | |
| AK146946.1 | 2 | 376 | BAE27555.1 | |
| AK166898.1 | 2 | 324 | BAE39101.1 | |
| AK166973.1 | 2 | 329 | BAE39156.1 | |
| AK168374.1 | 2 | 354 | BAE40306.1 | |
| BC012720.1 | 2 | 328 | AAH12720.1 | |
| D42083.1 | 2 | 269 | BAA07678.1 |
More Information
Set Membership: |
|

Genomic Map