Assay Details
Target Gene Details
Entrez Gene ID: | 26556 |
Gene Name: | homer homolog 1 (Drosophila) |
Gene Aliases: |
PSD-Zip45, SYN47, Ves-1, homer-1, vesl-1 |
Location: |
Chr.13:93304495-93405129 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Homer1 | NM_001284189.1 | 3 | 1032 | NP_001271118.1 |
| NM_011982.3 | 3 | 1032 | NP_036112.1 | |
| NM_147176.3 | 3 | 1032 | NP_671705.2 | |
| NM_152134.3 | 3 | 551 | NP_687036.1 | |
| AB019478.1 | 3 | 466 | BAA34353.1 | |
| AB019479.1 | 3 | 728 | BAA34354.1 | |
| AB089435.1 | 3 | 343 | BAC07257.1 | |
| AF093257.1 | 3 | 1034 | AAC71021.1 | |
| AF093258.1 | 3 | 381 | AAC71022.1 | |
| AY137385.1 | 3 | 551 | AAM95461.1 | |
| BC064041.1 | 3 | 852 | AAH64041.1 |

Genomic Map