Assay Details
Target Gene Details
Entrez Gene ID: | 16468 |
Gene Name: | jumonji, AT rich interactive domain 2 |
Gene Aliases: |
Jmj, jumonji |
Location: |
Chr.13:44729181-44921643 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jarid2 | NM_001205043.1 | 1 | 543 | NP_001191972.1 |
| NM_001205044.1 | 2 | 216 | NP_001191973.1 | |
| NM_021878.3 | 2 | 262 | NP_068678.1 | |
| AK044888.1 | 1 | 447 | ||
| AK045214.1 | 1 | 348 | BAC32264.1 | |
| AK147226.1 | 1 | 544 | BAE27780.1 | |
| BC052444.1 | 2 | 133 | AAH52444.1 | |
| BC060695.1 | 2 | 133 | AAH60695.1 | |
| D31967.1 | 2 | 152 | BAA06736.1 |
More Information
Set Membership: |
|

Genomic Map