Assay Details
Target Gene Details
Entrez Gene ID: | 57748 |
Gene Name: | junction-mediating and regulatory protein |
Gene Aliases: |
- |
Location: |
Chr.13:93430097-93499808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jmy | NM_021310.3 | NP_067285.2 | ||
| AB221690.1 | ||||
| AF201390.1 | AAF17555.1 | |||
| AK139934.1 | ||||
| AK141957.1 | BAE24898.1 | |||
| BC020052.1 | AAH20052.1 | |||
| BC069906.1 | AAH69906.1 | |||
| BC090835.1 | AAH90835.1 |

Genomic Map