Assay Details
Target Gene Details
Entrez Gene ID: | 18260 |
Gene Name: | occludin |
Gene Aliases: |
AI503564, Ocl |
Location: |
Chr.13:100496504-100553689 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ocln | NM_008756.2 | 5 | 1144 | NP_032782.1 |
| AK019880.1 | 5 | 1096 | BAB31900.1 | |
| AK033532.1 | 5 | 1077 | ||
| AK042576.1 | 5 | 1144 | BAC31298.1 | |
| AK145798.1 | 3 | 303 | BAE26656.1 | |
| BC138679.1 | 5 | 1127 | ||
| BC138680.1 | 5 | 1127 | ||
| BC145371.1 | 5 | 1142 | ||
| U49185.1 | 5 | 1187 | AAC52515.1 |
More Information
Set Membership: |
|

Genomic Map