Assay Details
Target Gene Details
Entrez Gene ID: | 12890 |
Gene Name: | complexin 2 |
Gene Aliases: |
921-L, AI413745, AW492120 |
Location: |
Chr.13:54371349-54383917 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cplx2 | NM_009946.3 | 1 | 152 | NP_034076.1 |
| CX241609.1 | 1 | 152 |

Genomic Map