Assay Details
Target Gene Details
Entrez Gene ID: | 218503 |
Gene Name: | FCH domain only 2 |
Gene Aliases: |
5832424M12Rik, AA387320 |
Location: |
Chr.13:98723406-98815769 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcho2 | NM_172591.3 | NP_766179.2 | ||
| AK031041.1 | BAC27226.1 | |||
| AK142282.1 | BAE25007.1 | |||
| AK146566.1 | BAE27264.1 | |||
| AK164135.1 | BAE37642.1 | |||
| BB620239.1 | ||||
| BC019933.1 | ||||
| BC052456.1 | AAH52456.1 | |||
| BC053718.1 | AAH53718.1 |
More Information
Set Membership: |
|

Genomic Map