Assay Details
Target Gene Details
Entrez Gene ID: | 30938 |
Gene Name: | FYVE, RhoGEF and PH domain containing 3 |
Gene Aliases: |
5830461L01Rik, ZFYVE5 |
Location: |
Chr.13:49263110-49320464 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgd3 | NM_015759.2 | 15 | 1845 | NP_056574.2 |
| AF017369.1 | 15 | 1880 | AAC35431.1 | |
| AK165413.1 | 15 | 2255 | BAE38171.1 | |
| BC132340.1 | 13 | 1664 | ||
| BC137954.1 | 13 | 1665 |
More Information
Set Membership: |
|

Genomic Map