Assay Details
Target Gene Details
Entrez Gene ID: | 17084 |
Gene Name: | lymphocyte antigen 86 |
Gene Aliases: |
MD-1, MD1 |
Location: |
Chr.13:37345345-37419036 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ly86 | NM_010745.2 | 1 | 123 | NP_034875.1 |
| AB007599.1 | 1 | 123 | BAA32399.1 | |
| AK027929.1 | 1 | 112 | BAC25670.1 | |
| BC065783.1 | AAH65783.1 | |||
| BC096414.1 | 1 | 91 | AAH96414.1 |

Genomic Map