Assay Details
Target Gene Details
Entrez Gene ID: | 18003 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated gene 9 |
Gene Aliases: |
Cas-L, CasL, HEF1, MEF1, Nedd-9, p105 |
Location: |
Chr.13:41309916-41487360 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nedd9 | NM_001111324.2 | 1 | 128 | NP_001104794.1 |
| AF009366.1 | 1 | 64 | AAB71663.1 | |
| AK030985.1 | 1 | 62 | BAC27203.1 | |
| AK083374.1 | 1 | 56 | BAC38890.1 | |
| BB842569.1 | 1 | 89 | ||
| BC053713.1 | 1 | 61 | AAH53713.1 | |
| BY001380.1 | 1 | 128 |

Genomic Map