Assay Details
Target Gene Details
Entrez Gene ID: | 18003 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated gene 9 |
Gene Aliases: |
Cas-L, CasL, HEF1, MEF1, Nedd-9, p105 |
Location: |
Chr.13:41309916-41487360 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nedd9 | NM_001111324.2 | NP_001104794.1 | ||
| NM_017464.5 | NP_059492.3 | |||
| AF009366.1 | AAB71663.1 | |||
| AK030985.1 | BAC27203.1 | |||
| AK033729.1 | BAC28451.1 | |||
| AK046357.1 | BAC32689.1 | |||
| AK054179.1 | BAC35682.1 | |||
| AK083374.1 | BAC38890.1 | |||
| BC053713.1 | AAH53713.1 |

Genomic Map