Assay Details
Target Gene Details
Entrez Gene ID: | 107513 |
Gene Name: | signal sequence receptor, alpha |
Gene Aliases: |
2510001K09Rik, 6330400D04, AI159733, AI452176, SSR, TRAPA |
Location: |
Chr.13:37971401-37994203 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssr1 | NM_025965.3 | NP_080241.3 | ||
| AB009295.1 | ||||
| AF326229.2 | AAK16151.2 | |||
| AF395811.1 | AAK82421.1 | |||
| AK010884.1 | BAB27245.1 | |||
| AK031538.1 | ||||
| AK160285.1 | BAE35725.1 | |||
| AK166235.1 | BAE38650.1 | |||
| AK167793.1 | BAE39823.1 | |||
| AK216988.1 | ||||
| BB619494.1 | ||||
| BC011255.1 | AAH11255.1 |

Genomic Map