Assay Details
Target Gene Details
Entrez Gene ID: | 75596 |
Gene Name: | prolactin family 7, subfamily b, member 1 |
Gene Aliases: |
1600014J19Rik, PLP-N, Prlpn |
Location: |
Chr.13:27601812-27610582 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl7b1 | NM_029355.2 | 5 | 540 | NP_083631.1 |
| AF525156.1 | 5 | 540 | AAN39704.1 | |
| AK005458.1 | 5 | 541 | BAB24049.1 | |
| BC116772.1 | 5 | 517 | ||
| BC119183.1 | 5 | 517 |
More Information
Set Membership: |
|

Genomic Map