Assay Details
Target Gene Details
Entrez Gene ID: | 20505 |
Gene Name: | solute carrier family 34 (sodium phosphate), member 1 |
Gene Aliases: |
NaPi-IIa, Npt2, Npt2a, Slc17a2 |
Location: |
Chr.13:55399622-55414695 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc34a1 | NM_011392.2 | NP_035522.2 | ||
| AB163922.1 | BAE48724.1 | |||
| AK018740.2 | BAB31381.1 | |||
| BC020529.1 | AAH20529.1 | |||
| L33878.1 | AAC42026.1 | |||
| U22465.1 | AAC52361.1 |
More Information
Set Membership: |
|

Genomic Map