Assay Details
Target Gene Details
Entrez Gene ID: | 12339 |
Gene Name: | calpain 7 |
Gene Aliases: |
AU022319, PalBH |
Location: |
Chr.14:31336620-31371991 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capn7 | NM_009796.2 | 7 | 943 | NP_033926.1 |
| AB028640.1 | 7 | 943 | BAA78731.1 | |
| AJ012475.1 | 7 | 756 | CAB39203.1 | |
| AK154277.1 | 7 | 1029 | ||
| AK169334.1 | 7 | 988 |

Genomic Map