Assay Details
Target Gene Details
Entrez Gene ID: | 218543 |
Gene Name: | splicing regulatory glutamine/lysine-rich protein 1 |
Gene Aliases: |
8430401B01, AI450757, AI462342, SRrp508, SRrp86, Sfrs12, Srsf12 |
Location: |
Chr.13:103739348-103774603 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srek1 | NM_172592.2 | 2 | 785 | NP_766180.1 |
| AK033354.1 | 2 | 785 | BAC28243.1 | |
| AK037115.1 | 2 | 761 | BAC29709.2 | |
| AK039311.1 | 4 | 520 | BAC30314.1 | |
| AK044086.1 | 1 | 89 | BAC31767.1 | |
| AK052265.1 | 4 | 654 | BAC34905.2 | |
| AK054414.1 | 2 | 1236 | ||
| BC063761.1 | 5 | 530 | AAH63761.1 | |
| BC070460.1 | 2 | 158 | AAH70460.1 |
More Information
Set Membership: |
|

Genomic Map