Assay Details
Target Gene Details
Entrez Gene ID: | 20963 |
Gene Name: | spleen tyrosine kinase |
Gene Aliases: |
Sykb |
Location: |
Chr.13:52582329-52648792 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syk | NM_001198977.1 | 11 | 1636 | NP_001185906.1 |
| NM_011518.2 | 11 | 1640 | NP_035648.2 | |
| AK049207.1 | 11 | 1665 | ||
| AK136363.1 | 11 | 1639 | BAE22946.1 | |
| AK143564.1 | 11 | 1669 | BAE25438.1 | |
| BC065121.1 | 11 | 1572 | AAH65121.1 | |
| U25685.1 | 10 | 1424 | AAA87462.1 | |
| U36776.1 | 11 | 1643 | AAA79996.1 | |
| Z49877.1 | 11 | 1898 | CAA90034.1 |

Genomic Map