Assay Details
Target Gene Details
Entrez Gene ID: | 320234 |
Gene Name: | coiled-coil domain containing 66 |
Gene Aliases: |
E230015L20Rik |
Location: |
Chr.14:27481090-27508506 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc66 | NM_177111.3 | 18 | 2812 | NP_796085.3 |
| AK053185.1 | 3 | 362 | BAC35302.1 | |
| AK087583.1 | 10 | 1533 | BAC39936.1 | |
| AK158666.1 | 16 | 2330 | BAE34603.1 | |
| AK170006.1 | 16 | 2279 | ||
| BC070471.1 | 18 | 2813 | ||
| BC072583.1 | 10 | 1929 | AAH72583.1 |
Target Gene Details
Entrez Gene ID: | 218850 |
Gene Name: | family with sequence similarity 208, member A |
Gene Aliases: |
4732418E12, 4933409E02Rik, D14Abb1e |
Location: |
Chr.14:27426658-27483555 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam208a | NM_001114879.1 | 24 | 6288 | NP_001108351.1 |
| NM_028945.1 | 24 | 6562 | NP_083221.1 | |
| AK173099.1 | 4 | 2023 | BAD32377.1 |

Genomic Map