Assay Details
Target Gene Details
Entrez Gene ID: | 67310 |
Gene Name: | prolactin family8, subfamily a, member 9 |
Gene Aliases: |
1600019F10Rik, PLP-Cbeta, Prlpc2 |
Location: |
Chr.13:27558001-27564616 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl8a9 | NM_023332.3 | NP_075821.1 | ||
| AF158744.1 | AAK81734.1 | |||
| AK005409.1 | BAB24008.1 | |||
| AK005494.1 | BAB24079.1 | |||
| BC064762.1 | AAH64762.1 |
More Information
Set Membership: |
|

Genomic Map