Assay Details
Target Gene Details
Entrez Gene ID: | 51791 |
Gene Name: | regulator of G-protein signaling 14 |
Gene Aliases: |
RPIP1 |
Location: |
Chr.13:55369712-55384691 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs14 | NM_016758.3 | NP_058038.2 | ||
| AK002891.1 | BAB22436.1 | |||
| AK154944.1 | ||||
| BC030321.1 | AAH30321.1 | |||
| U85055.1 | AAB41893.1 |

Genomic Map