Assay Details
Target Gene Details
Entrez Gene ID: | 218543 |
Gene Name: | splicing regulatory glutamine/lysine-rich protein 1 |
Gene Aliases: |
8430401B01, AI450757, AI462342, SRrp508, SRrp86, Sfrs12, Srsf12 |
Location: |
Chr.13:103739348-103774603 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srek1 | NM_172592.2 | NP_766180.1 | ||
| AK033354.1 | BAC28243.1 | |||
| AK075652.1 | ||||
| BC070460.1 | AAH70460.1 |
More Information
Set Membership: |
|

Genomic Map