Assay Details
Target Gene Details
Entrez Gene ID: | 70484 |
Gene Name: | solute carrier family 35, member D2 |
Gene Aliases: |
5730408I21Rik, SQV7L, UGTrel8, hfrc, mUGTrel8 |
Location: |
Chr.13:64096308-64129381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35d2 | NM_001001321.3 | NP_001001321.2 | ||
| AB117931.1 | BAD18884.1 | |||
| AB117932.1 | BAD18885.1 | |||
| AK154257.1 | BAE32469.1 | |||
| AK159148.1 | BAE34855.1 | |||
| BC046402.1 | AAH46402.1 | |||
| BC107210.1 | AAI07211.1 | |||
| BC107211.1 | AAI07212.1 |
More Information
Set Membership: |
|

Genomic Map