Assay Details
Target Gene Details
Entrez Gene ID: | 20963 |
Gene Name: | spleen tyrosine kinase |
Gene Aliases: |
Sykb |
Location: |
Chr.13:52582329-52648792 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syk | NM_001198977.1 | 9 | 1323 | NP_001185906.1 |
| NM_011518.2 | 9 | 1327 | NP_035648.2 | |
| AK049207.1 | 9 | 1352 | ||
| AK136363.1 | 9 | 1326 | BAE22946.1 | |
| AK143564.1 | 9 | 1356 | BAE25438.1 | |
| BC065121.1 | 9 | 1259 | AAH65121.1 | |
| U25685.1 | 8 | 1111 | AAA87462.1 | |
| U36776.1 | 9 | 1330 | AAA79996.1 | |
| Z49877.1 | 9 | 1585 | CAA90034.1 |

Genomic Map