Assay Details
Target Gene Details
Entrez Gene ID: | 67046 |
Gene Name: | TBC1 domain family, member 7 |
Gene Aliases: |
2610009C09Rik |
Location: |
Chr.13:43151740-43171501 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d7 | NM_001252639.1 | 1 | 81 | NP_001239568.1 |
| NM_001252640.1 | 1 | 81 | NP_001239569.1 | |
| NM_025935.3 | 1 | 81 | NP_080211.1 | |
| AK011356.1 | BAB27564.1 | |||
| AK049036.1 | 1 | 51 | BAC33521.1 | |
| AK156543.1 | BAE33751.1 | |||
| BY166141.1 | 1 | 82 |
More Information
Set Membership: |
|

Genomic Map