Assay Details
Target Gene Details
Entrez Gene ID: | 74257 |
Gene Name: | tetraspanin 17 |
Gene Aliases: |
2210021G21Rik, AI047581, Fbxo23, Tm4sf17 |
Location: |
Chr.13:54789352-54796896 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan17 | NM_028841.3 | NP_083117.2 | ||
| AK008761.1 | BAB25880.1 | |||
| AK166272.1 | BAE38673.1 | |||
| AK171691.1 | BAE42615.1 | |||
| BC010346.1 | AAH10346.1 | |||
| BY734818.1 |

Genomic Map