Assay Details
Target Gene Details
Entrez Gene ID: | 19134 |
Gene Name: | PRP4 pre-mRNA processing factor 4 homolog B (yeast) |
Gene Aliases: |
2610037H07, Prp4, Prp4k, Prpk, cbp143 |
Location: |
Chr.13:34875296-34906064 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf4b | NM_013830.2 | NP_038858.2 | ||
| AB516304.1 | ||||
| AF033663.1 | AAC32042.1 | |||
| AF283466.1 | AAM19102.1 | |||
| AK076020.1 | BAC36125.1 | |||
| AK083926.1 | BAC39069.1 | |||
| BC141272.1 | ||||
| BC141273.1 | ||||
| U48737.1 | AAB03269.1 |

Genomic Map